Recombinant HERV-K (from E. coli)
Vial
Catalog # 77682-828
Supplier: Creative Biomart
Supplier #: SARS-402
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Recombinant HERV-K truncated protein, his-tagged.
Specifications
Specifications
- Pack type:Vial
- Cat. No.:77682-828
- Molecular Weight:51.5 kDa
- Shipping Temperature:Dry Ice
- Protein/Peptide Type:Recombinant
- Formulation:10 mM Tris-HCl, pH 7.2, 8 M urea and 2 mM EDTA.
- Endotoxin Level:Very low
- Purity:Protein is >90% pure as determined by SDS PAGE.
- Protein Synonyms:HERV-K
- Protein/Peptide Name:HERV-K
- Source:E. coli
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



