SARS Recombinant Envelope protein
Catalog # 77682-812
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:SARS
- Protein Synonyms:ORF4|structural protein|E protein|E|envelope protein
- Protein/Peptide Name:Envelope protein
- Purity:SARS Envelope protein is >95% pure as determined by 10% PAGE(coomassie staining).
- Molecular Weight:34 kDa
- Endotoxin Level:Very low
- Formulation:50 mM Tris-HCl, 60 mM NaCl, pH 8 and 50% glycerol.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-812
Specifications
About this item
Recombinant SARS Envelope Protein.