Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antibodies
BNT162c2 SARS-CoV-2 Vaccine
BNT162c2 SARS-CoV-2 Vaccine
BNT162c2 SARS-CoV-2 Vaccine
Catalog # 77682-744
Supplier:
Creative Biomart
New Product
Expand image
BNT162c2 SARS-CoV-2 Vaccine
Catalog # 77682-744
Supplier:
Creative Biomart
Supplier Number:
INB-326
New Product
Specifications
Reactivity:
SARS-CoV-2
Shipping Temperature:
Dry Ice
Cat. No.:
77682-744
Packaging:
Vial
Specifications
About this item
Product Details
Product Details