Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
SARS Recombinant Matrix protein

SARS Recombinant Matrix protein

Vial
Catalog # 77682-632
Supplier:   Creative Biomart
Supplier #:  HIV-275
SARS Recombinant Matrix protein
Catalog # 77682-632
Supplier:  Creative Biomart
CAS Number:  
SARS Recombinant Matrix protein
SARS Recombinant Matrix protein

Vial

CAS Number:  
Price
$1,022.60
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recombinant SARS Matrix antigen.

Specifications

Specifications

  • Pack type:
    Vial
  • Cat. No.:
    77682-632
  • Molecular Weight:
    30 kDa
  • Shipping Temperature:
    Dry Ice
  • Protein/Peptide Type:
    Recombinant
  • Formulation:
    50 mM Tris-HCl, 60 mM NaCl, and 50% glycerol.
  • Endotoxin Level:
    Very low
  • Purity:
    SARS Matrix Protein is >95% pure as determined by 10% PAGE (coomassie staining).
  • Protein Synonyms:
    Matrix protein|SARS
  • Protein/Peptide Name:
    Matrix protein
  • Species:
    SARS

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.