Anti-TP53BP2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TP53BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77706-591
- 77706-590
- Antigen Symbol:
- TP53BP2
- TP53BP2
- Antigen Synonyms:
- ASPP2|PPP1R13A|Tumor suppressor p53 binding protein 2|ASPP2_HUMAN|Tumor protein p53 binding protein 2|p53BP2|Tp53bp2|Apoptosis-stimulating of p53 protein 2|p53 binding protein 2|NY REN 51 antigen |53BP2|Bcl2-binding protein|BBP|Tumor suppressor p53-binding protein 2|Renal carcinoma antigen NY-REN-51|Apoptosis stimulating protein of p53 2|Bcl2 binding protein|Apoptosis stimulating of p53 protein 2|p53-binding protein 2
- ASPP2|PPP1R13A|Tumor suppressor p53 binding protein 2|ASPP2_HUMAN|Tumor protein p53 binding protein 2|p53BP2|Tp53bp2|Apoptosis-stimulating of p53 protein 2|p53 binding protein 2|NY REN 51 antigen |53BP2|Bcl2-binding protein|BBP|Tumor suppressor p53-binding protein 2|Renal carcinoma antigen NY-REN-51|Apoptosis stimulating protein of p53 2|Bcl2 binding protein|Apoptosis stimulating of p53 protein 2|p53-binding protein 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13625
- Q13625
- Isotype:
- IgG
- IgG
- Immunogen:
- Synthetic peptide of Human TP53BP2
- Synthetic peptide of Human TP53BP2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice