Anti-CNTROB Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, WB:1:200-1:1000
Type:
Antigen: CNTROB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77706-102
- 77706-103
- Antigen Symbol:
- CNTROB
- CNTROB
- Antigen Synonyms:
- LYST-interacting protein LIP8|Centrosomal BRCA2-interacting protein|centrosomal BRCA2 interacting protein|RP23 26L6.9|CNTRB_HUMAN|9830165K03Rik|Centrobin|Cntrob|RGD1307488|LYST-interacting protein 8|PP1221|LYST interacting protein 8|LIP8
- LYST-interacting protein LIP8|Centrosomal BRCA2-interacting protein|centrosomal BRCA2 interacting protein|RP23 26L6.9|CNTRB_HUMAN|9830165K03Rik|Centrobin|Cntrob|RGD1307488|LYST-interacting protein 8|PP1221|LYST interacting protein 8|LIP8
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N137
- Q8N137
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CNTROB
- Fusion protein of Human CNTROB
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice