Anti-CNTN4 Rabbit Polyclonal Antibody
Catalog # 77701-987
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CNTN4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q8IWV2
- Antigen Synonyms:Brain-derived immunoglobulin superfamily protein 2|Cntn4|Neural cell adhesion protein BIG 2|BIG 2|Contactin-4|SCA16|AXCAM|CNTN4A|contactin 4|Axonal associated cell adhesion molecule|Brain derived immunoglobulin superfamily protein 2|BIG-2|CNTN4_HUMAN
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Contactin 4.
- Purification:Antigen affinity purified
- Cat. No.:77701-987
- Packaging:Vial
Specifications
About this item
IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: CNTN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat