Anti-MOB3C Rabbit Polyclonal Antibody
Catalog # 77701-959
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MOB3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q70IA8
- Antigen Synonyms:Mps one binder kinase activator-like 2C|MOB kinase activator 3C|Mob1 homolog 2C|MOBKL2C|MOB3C
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Mob3C.
- Purification:Antigen affinity purified
- Cat. No.:77701-959
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: MOB3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse