Anti-CSNK1A1/CSNK1A1L (phospho-Y294) Rabbit Polyclonal Antibody
77701-931
New Product
- Antigen Symbol:CSNK1A1/CSNK1A1L (phospho-Y294)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P48729/Q8N752
- Antigen Synonyms:CKI-alpha|CSNK1A1L|Casein kinase I isoform alpha|CK1|CSNK1A1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Casein Kinase Iα around the phosphorylation site of Y294.
- Purification:Antigen affinity purified
- Cat. No.:77701-931
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: CSNK1A1/CSNK1A1L (phospho-Y294)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat