Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-COL13A1 Rabbit Polyclonal Antibody
Anti-COL13A1 Rabbit Polyclonal Antibody
Catalog # 77701-833
Supplier:  AFG Bioscience
New Product
undefined
Anti-COL13A1 Rabbit Polyclonal Antibody
Catalog # 77701-833
Supplier:  AFG Bioscience
Supplier Number:  A19797-100UG
New Product

Specifications

  • Antigen Symbol:
    COL13A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    Q5TAT6
  • Antigen Synonyms:
    collagen type XIII alpha 1|collagen alpha 1(XIII) chain |CODA1_HUMAN|COLXIIIA1|Collagen alpha-1(XIII) chain|COL13A1
  • Storage Buffer:
    PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
  • Storage Temperature:
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    Synthesized peptide derived from the C-terminal region of Human COL13A1.
  • Purification:
    Antigen affinity purified
  • Cat. No.:
    77701-833
  • Packaging:
    Vial

Specifications

About this item

IF:1:200-1:1000, ELISA:1:20000

Type:
Antigen: COL13A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse