Anti-OR4B1 Rabbit Polyclonal Antibody
77701-759
New Product
- Antigen Symbol:OR4B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q8NGF8
- Antigen Synonyms:OR4B1|Olfactory receptor 4B1|OST208|Olfactory receptor OR11-106
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human Olfactory receptor 4B1.
- Purification:Antigen affinity purified
- Cat. No.:77701-759
- Packaging:Vial
IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: OR4B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human