Anti-POLI Rabbit Polyclonal Antibody
Catalog # 77701-696
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:POLI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9UNA4
- Antigen Synonyms:RAD30B|Eta 2|RAD3OB|RAD30, S. cerevisiae, homolog of|Eta2|RAD 30B|DNA polymerase iota|POLI_HUMAN|POLI|Polymerase (DNA directed) iota|RAD30 homolog B
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human DNA pol ι.
- Purification:Antigen affinity purified
- Cat. No.:77701-696
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: POLI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human