Anti-SLC27A4 Rabbit Polyclonal Antibody
Catalog # 77701-660
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SLC27A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q6P1M0
- Antigen Synonyms:Long-chain fatty acid transport protein 4|FATP-4|ACSVL4|Fatty acid transport protein 4|FATP4|SLC27A4|Solute carrier family 27 member 4|EC 6.2.1.-
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human ACSVL4.
- Purification:Antigen affinity purified
- Cat. No.:77701-660
- Packaging:Vial
Specifications
About this item
IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: SLC27A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human