Anti-CLDN20 Rabbit Polyclonal Antibody
Catalog # 77687-409
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CLDN20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P56880
- Antigen Synonyms:CLD20_HUMAN|CLDN20|Claudin-20
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Claudin-20 protein (29-81AA)
- Purification:>95%, Protein G purified
- Cat. No.:77687-409
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000, IF:1:50-1:200
Type:
Antigen: CLDN20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human