Anti-UBE2S Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:200-1:1000, IHC:1:50-1:200
Type:
Antigen: UBE2S
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77709-715
- 77709-714
- Antigen Synonyms:
- Ubiquitin-conjugating enzyme E2-EPF5|E2 EPF|E2-EPF|Ubiquitin conjugating enzyme E2S|Ubiquitin carrier protein S|E2EPF|UBE2S_HUMAN|EPF5|Ubiquitin conjugating enzyme E2 24 kDa|Ubiquitin conjugating enzyme E2 24 kD|AA409170|Ube2s|Ubiquitin conjugating enzyme E2 S|Ubiquitin-protein ligase S|Ubiquitin-conjugating enzyme E2-24 kDa|Ubiquitin-conjugating enzyme E2 S|Ubiquitin conjugating enzyme E2 EPF5|Ubiquitin protein ligase S|MGC101982
- Ubiquitin-conjugating enzyme E2-EPF5|E2 EPF|E2-EPF|Ubiquitin conjugating enzyme E2S|Ubiquitin carrier protein S|E2EPF|UBE2S_HUMAN|EPF5|Ubiquitin conjugating enzyme E2 24 kDa|Ubiquitin conjugating enzyme E2 24 kD|AA409170|Ube2s|Ubiquitin conjugating enzyme E2 S|Ubiquitin-protein ligase S|Ubiquitin-conjugating enzyme E2-24 kDa|Ubiquitin-conjugating enzyme E2 S|Ubiquitin conjugating enzyme E2 EPF5|Ubiquitin protein ligase S|MGC101982
- Gene ID:
- Q16763
- Q16763
- Immunogen:
- Fusion protein of Human UBE2S
- Fusion protein of Human UBE2S
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice