Anti-BAZ1A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, IHC:1:25-1:100
Type:
Antigen: BAZ1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77709-625
- 77709-624
- Antigen Synonyms:
- Williams syndrome transcription factor-related chromatin-remodeling factor 180|FLJ14383|drosophila|Baz1a|DKFZp586E0518|CHRAC subunit ACF1|Gtl5|hACF1|WALp1|hWALp1|Williams syndrome transcription factor related chromatin remodeling factor 180|Bromodomain adjacent to zinc finger domain 1A|BC065123|B930060C03|BAZ1A_HUMAN|ATP utilizing chromatin assembly and remodeling factor 1|Bromodomain adjacent to zinc finger domain protein 1A|ATP-utilizing chromatin assembly and remodeling factor 1|homolog of|cbp146|ATP-dependent chromatin-remodeling protein|WCRF180|ATP dependent chromatin remodelling protein|Acf1
- Williams syndrome transcription factor-related chromatin-remodeling factor 180|FLJ14383|drosophila|Baz1a|DKFZp586E0518|CHRAC subunit ACF1|Gtl5|hACF1|WALp1|hWALp1|Williams syndrome transcription factor related chromatin remodeling factor 180|Bromodomain adjacent to zinc finger domain 1A|BC065123|B930060C03|BAZ1A_HUMAN|ATP utilizing chromatin assembly and remodeling factor 1|Bromodomain adjacent to zinc finger domain protein 1A|ATP-utilizing chromatin assembly and remodeling factor 1|homolog of|cbp146|ATP-dependent chromatin-remodeling protein|WCRF180|ATP dependent chromatin remodelling protein|Acf1
- Gene ID:
- Q9NRL2
- Q9NRL2
- Immunogen:
- Synthetic peptide of Human BAZ1A
- Synthetic peptide of Human BAZ1A
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice