Anti-TRIM27 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRIM27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77709-222
- 77709-221
- Antigen Synonyms:
- MGC189472|DAAP-182E11.3|Rfp1|RNF76|Tripartite motif containing protein 27|AW538890|RING finger protein 76|RFP transforming protein|TRIM 27|Tripartite motif containing 27|Tripartite motif-containing protein 27|Tripartite motif protein TRIM27|OTTHUMP00000029314|Ret finger protein|RFP|TRI27_HUMAN|Trim27|Zinc finger protein RFP
- MGC189472|DAAP-182E11.3|Rfp1|RNF76|Tripartite motif containing protein 27|AW538890|RING finger protein 76|RFP transforming protein|TRIM 27|Tripartite motif containing 27|Tripartite motif-containing protein 27|Tripartite motif protein TRIM27|OTTHUMP00000029314|Ret finger protein|RFP|TRI27_HUMAN|Trim27|Zinc finger protein RFP
- Gene ID:
- P14373
- P14373
- Immunogen:
- Synthetic peptide of Human TRIM27
- Synthetic peptide of Human TRIM27
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice