Anti-NME2 Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:10000, WB:1:1000-1:5000, IHC:1:30-1:150
Type:
Antigen: NME2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77708-891
- 77708-890
- Antigen Synonyms:
- NDP kinase B|C myc purine binding transcription factor PUF|epididymis secretory sperm binding protein Li 155an|C-myc purine-binding transcription factor PUF|Non metastatic cells 2|HEL-S-155an|non-metastatic cells 2|NM23 H2|protein (NM23B) expressed in|Histidine protein kinase NDKB|Nucleotide diphosphate kinase B|NME/NM23 nucleoside diphosphate kinase 2|C myc transcription factor|NDKB_HUMAN|NDPK B|nm23-H2|NDK B|protein (NM23) expressed in|PUF|NDKB|NDPKB|MGC111212|MGC2212|nme2|NM23B|Nucleoside diphosphate kinase B
- NDP kinase B|C myc purine binding transcription factor PUF|epididymis secretory sperm binding protein Li 155an|C-myc purine-binding transcription factor PUF|Non metastatic cells 2|HEL-S-155an|non-metastatic cells 2|NM23 H2|protein (NM23B) expressed in|Histidine protein kinase NDKB|Nucleotide diphosphate kinase B|NME/NM23 nucleoside diphosphate kinase 2|C myc transcription factor|NDKB_HUMAN|NDPK B|nm23-H2|NDK B|protein (NM23) expressed in|PUF|NDKB|NDPKB|MGC111212|MGC2212|nme2|NM23B|Nucleoside diphosphate kinase B
- Gene ID:
- P22392
- P22392
- Immunogen:
- Synthetic peptide of Human NME2
- Synthetic peptide of Human NME2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice