Anti-ZP1 Rabbit Polyclonal Antibody
77702-332
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Host:
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P60852
- Antigen Synonyms:Zp-1|Zp 1|Zona pellucida glycoprotein 1|zona pellucida glycoprotein 1 (sperm receptor)|MGC87693|zona pellucida protein 1|ZP1_HUMAN|Processed zona pellucida sperm-binding protein 1|Zp1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human ZP1.
- Purification:Antigen affinity purified
- Cat. No.:77702-332
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: ZP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human