Anti-ESR1 (phospho-S305) Rabbit Polyclonal Antibody
Catalog # 77702-237
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:P03372
- Antigen Synonyms:Estradiol receptor|Estrogen receptor alpha delta 3*,4,5,6,7*/819 2 isoform|8*/901 isoform|ER|Era|ER alpha|Estrogen receptor alpha|Estrogen receptor alpha 3*,4,5,6,7*/822 isoform|Estrogen receptor alpha delta 4*,5,6,7*/654 isoform|7*/654 isoform|ESRA|NR3A1|Estrogen receptor|Estrogen receptor alpha E1 N2 E2 1 2|ER-alpha|Estrogen receptor alpha E1 E2 1 2|ESR|ESR1|Nuclear receptor subfamily 3 group A member 1|Estrogen receptor 1|Estrogen nuclear receptor alpha|DKFZp686N23123|ESTRR|Estrogen receptor alpha delta 3*,4,5,6,7*,8*/941 isoform|Estrogen receptor alpha delta 4 +49 isoform|ESR1_HUMAN|Estrogen receptor alpha delta 4*,5,6,7,8*/901 isoform|7*/822 isoform|7*/819 2 isoform|8*/941 isoform
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human ERα around the phosphorylation site of S305.
- Purification:Antigen affinity purified
- Cat. No.:77702-237
- Packaging:Vial
Specifications
About this item
IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: ESR1 (phospho-S305)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human