Anti-ERBB2 Rabbit Polyclonal Antibody
77702-212
New Product
- Form:Liquid
- Gene ID:P04626
- Antigen Synonyms:Human epidermal growth factor receptor 2|NEU|V erb b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian)|MLN19|Tyrosine kinase-type cell surface receptor HER2|Cerb B2/neu protein|Neuroblastoma/glioblastoma derived oncogene homolog|TKR1|C erb B2/neu protein|Neuro/glioblastoma derived oncogene homolog|V erb b2 avian erythroblastic leukemia viral oncogene homolog 2 (neuro/glioblastoma derived oncogene homolog)|Erb b2 receptor tyrosine kinase 2|ERBB2|HER 2/NEU|MLN 19|Herstatin|CD340|V erb b2 avian erythroblastic leukemia viral oncogene homolog 2|NEU proto oncogene|ERBB2_HUMAN|CD340 antigen|HER2|Metastatic lymph node gene 19 protein|HER 2|Verb b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian)|V erb b2 avian erythroblastic leukemia viral oncoprotein 2|p185erbB2|Receptor tyrosine-protein kinase erbB-2|Proto-oncogene c-ErbB-2|V erb b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog|Tyrosine kinase type cell surface receptor HER2|ErbB-2 proto-oncogene|Proto-oncogene Neu|CerbB2|Verb b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog|NGL
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Neu around the non-phosphorylation site of T686.
- Purification:Antigen affinity purified
- Cat. No.:77702-212
- Packaging:Vial
IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat