Anti-AHR/AHRR (phospho-S36) Rabbit Polyclonal Antibody
77702-135
New Product
- Form:Liquid
- Gene ID:P35869/A9YTQ3
- Antigen Synonyms:AhR repressor|BHLHE76|AHRR|bHL|Class E basic helix-loop-helix protein 77|KIAA1234|AHR|BHLHE77|Ah receptor|Aryl hydrocarbon receptor repressor|Class E basic helix-loop-helix protein 76|Aryl hydrocarbon receptor
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Ah Receptor around the phosphorylation site of S36.
- Purification:Antigen affinity purified
- Cat. No.:77702-135
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: AHR/AHRR (phospho-S36)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat