Polysulfide Oil Standard: 500 µg/g S in Mineral Oil, VHG, LGC Standards
Catalog # 77697-821
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:13 cSt Mineral Oil
- Description:Polysulfide Oil Standard: 500 µg/g S in Mineral Oil
- Size:100 mL
- Cat. No.:77697-821