Matrix Blank - 15W40 Motor Oil with no Diesel Added, Bottled in a 100mL Wide Mouth Glass Bottle for Use with GC or IR., VHG, LGC Standards
Catalog # 77697-513
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Oil
- Description:Matrix Blank - 15W40 Motor Oil with no Diesel Added, Bottled in a 100mL Wide Mouth Glass Bottle for Use with GC or IR.
- Size:100 mL
- Cat. No.:77697-513