Cold Filter Plugging Point, Diesel (Nominal: -21.7°C), Paragon Scientific, LGC Standards
Catalog # 77691-398
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Cold Filter Plugging Point, Diesel (Nominal: -21.7°C)
- Size:250 mL
- Cat. No.:77691-398