Cleveland Open Cup (Nominal: 161.4 °C), Flash Point, Paragon Scientific, LGC Standards
Catalog # 77691-400
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Cleveland Open Cup (Nominal: 161.4 °C), Flash Point
- Size:3 x 80 ml
- Cat. No.:77691-400