Order Entry
Puerto Rico
ContactUsLinkComponent
PMCC Procedure B, Lubricant (Nominal: 100.7°C), Flash Point, Paragon Scientific, LGC Standards
PMCC Procedure B, Lubricant (Nominal: 100.7°C), Flash Point, Paragon Scientific, LGC Standards
Catalog # 77696-023
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
PMCC Procedure B, Lubricant (Nominal: 100.7°C), Flash Point, Paragon Scientific, LGC Standards
Catalog # 77696-023
Supplier:  LGC Standards
Supplier Number:  ALK-CRMU-PMLUB
New Product
CAS Number:  

Specifications

  • Description:
    PMCC Procedure B, Lubricant (Nominal: 100.7°C), Flash Point
  • Size:
    250 mL
  • Cat. No.:
    77696-023

Specifications

About this item