Pour Point, Lubricant (Nominal: -26.1°C), Paragon Scientific, LGC Standards
Catalog # 77696-026
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Pour Point, Lubricant (Nominal: -26.1°C)
- Size:250 mL
- Cat. No.:77696-026