TAG, Jet Aviation Fuel (Nominal: 40.4°C), Flash Point, Paragon Scientific, LGC Standards
Catalog # 77693-348
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:TAG, Jet Aviation Fuel (Nominal: 40.4°C), Flash Point
- Size:250 mL
- Cat. No.:77693-348