Density 15°C (Nominal: 0.8494 g/mL @ 15°C), Paragon Scientific, LGC Standards
Catalog # 77691-407
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:Density 15°C (Nominal: 0.8494 g/mL @ 15°C)
- Size:60 ml
- Cat. No.:77691-407