Order Entry
Puerto Rico
ContactUsLinkComponent
Density 20°C (Nominal: 0.8723 g/mL @ 20°C), Paragon Scientific, LGC Standards
Density 20°C (Nominal: 0.8723 g/mL @ 20°C), Paragon Scientific, LGC Standards
Catalog # 77691-408
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Density 20°C (Nominal: 0.8723 g/mL @ 20°C), Paragon Scientific, LGC Standards
Catalog # 77691-408
Supplier:  LGC Standards
Supplier Number:  ALK-DEN20-06
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Description:
    Density 20°C (Nominal: 0.8723 g/mL @ 20°C)
  • Size:
    60 ml
  • Cat. No.:
    77691-408

Specifications

About this item