Density 20°C (Nominal: 1.6218 g/mL @ 20°C), Paragon Scientific, LGC Standards
Catalog # 77691-409
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:Density 20°C (Nominal: 1.6218 g/mL @ 20°C)
- Size:60 ml
- Cat. No.:77691-409