Density 25°C (Nominal: 0.9797 g/mL @ 25°C), Paragon Scientific, LGC Standards
Catalog # 77691-410
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:Density 25°C (Nominal: 0.9797 g/mL @ 25°C)
- Size:60 ml
- Cat. No.:77691-410