Order Entry
Puerto Rico
ContactUsLinkComponent
Pure Water Density (Nominal: 0.99909 g/mL @ 15°C), Values at 15, 20 & 25°C, Paragon Scientific, LGC Standards
Pure Water Density (Nominal: 0.99909 g/mL @ 15°C), Values at 15, 20 & 25°C, Paragon Scientific, LGC Standards
Catalog # 77696-030
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Pure Water Density (Nominal: 0.99909 g/mL @ 15°C), Values at 15, 20 & 25°C, Paragon Scientific, LGC Standards
Catalog # 77696-030
Supplier:  LGC Standards
Supplier Number:  ALK-DEN-WAT3
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Description:
    Pure Water Density (Nominal: 0.99909 g/mL @ 15°C), Values at 15, 20 & 25°C
  • Size:
    3 x 60 ml
  • Cat. No.:
    77696-030

Specifications

About this item