Pensky Martens (Nominal: 137.5°C), Flash Point, Paragon Scientific, LGC Standards
Catalog # 77696-035
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Pensky Martens (Nominal: 137.5°C), Flash Point
- Size:3 x 80 ml
- Cat. No.:77696-035