Order Entry
Puerto Rico
ContactUsLinkComponent
Type JF1-L (Certified @ -20 & -40°C), Low Temperature Viscosity, Paragon Scientific, LGC Standards
Type JF1-L (Certified @ -20 & -40°C), Low Temperature Viscosity, Paragon Scientific, LGC Standards
Catalog # 77693-357
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Type JF1-L (Certified @ -20 & -40°C), Low Temperature Viscosity, Paragon Scientific, LGC Standards
Catalog # 77693-357
Supplier:  LGC Standards
Supplier Number:  ALK-JF1-L
New Product
CAS Number:  

Specifications

  • Description:
    Type JF1-L (Certified @ -20 & -40°C), Low Temperature Viscosity
  • Size:
    500 ml
  • Cat. No.:
    77693-357

Specifications

About this item