Order Entry
Puerto Rico
ContactUsLinkComponent
ASTM D5453 Curve 1, S @ 0.5 -10ppm in isooctane, VHG, LGC Standards
ASTM D5453 Curve 1, S @ 0.5 -10ppm in isooctane, VHG, LGC Standards
Catalog # 77691-417
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
ASTM D5453 Curve 1, S @ 0.5 -10ppm in isooctane, VHG, LGC Standards
Catalog # 77691-417
Supplier:  LGC Standards
Supplier Number:  ALK-KTSISOV6-CRV1
New Product
CAS Number:  

Specifications

  • Matrix:
    Isooctane
  • Description:
    ASTM D5453 Curve 1, S @ 0.5 -10ppm in isooctane
  • Size:
    6 x 2 mL
  • Cat. No.:
    77691-417

Specifications

About this item