Sulfur Calibration Set: S @ 25-1000 mg/Kg in Synthetic Diesel, VHG, LGC Standards
Catalog # 77693-360
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Synthetic Diesel
- Description:Sulfur Calibration Set: S @ 25-1000 mg/Kg in Synthetic Diesel
- Size:5 x 2 mL
- Cat. No.:77693-360