DRO Mixture 2000 µg/mL in Dichloromethane/Hexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-348
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Dichloromethane:Hexane
- Description:DRO Mixture 2000 µg/mL in Dichloromethane/Hexane
- Size:1 mL
- Cat. No.:77677-348