Oregon Pesticide Mixture 1-100 AL 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-374
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Acetonitrile
- Description:Oregon Pesticide Mixture 1-100 AL 100 µg/mL in Acetonitrile
- Size:1 mL
- Cat. No.:77696-374