ASTM Method D3606 Check Standard 10-100 mL/L in Isooctane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-740
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Isooctane
- Description:ASTM Method D3606 Check Standard 10-100 mL/L in Isooctane
- Size:10 x 2 mL
- Cat. No.:77691-740