ASTM Method D5769 Quality Control Reference Material with 3 IS in Isooctane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-741
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Isooctane
- Description:ASTM Method D5769 Quality Control Reference Material with 3 IS in Isooctane
- Size:10 x 2 mL
- Cat. No.:77691-741