ASTM Method D5501 96% Ethanol QC Check Mixture in Ethanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-783
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Ethanol
- Description:ASTM Method D5501 96% Ethanol QC Check Mixture in Ethanol
- Size:10 x 2 mL
- Cat. No.:77691-783