ASTM Method D3606 Benzene in Gasoline Kit with 10% EtOH & IS in Isooctane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-786
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Isooctane
- Description:ASTM Method D3606 Benzene in Gasoline Kit with 10% EtOH & IS in Isooctane
- Size:7 x 2 mL
- Cat. No.:77691-786