ASTM Method D4815 Oxygenates in Gasoline Calibration Kit with IS in Oxygenate Free Gasoline, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-787
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Oxygenate Free Gasoline
- Description:ASTM Method D4815 Oxygenates in Gasoline Calibration Kit with IS in Oxygenate Free Gasoline
- Size:KIT
- Cat. No.:77691-787