Oregon Cannabis Solvent Mixture 1000 µg/mL in Triacetin, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-421
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Triacetin
- Description:Oregon Cannabis Solvent Mixture 1000 µg/mL in Triacetin
- Size:5 x 1 mL
- Cat. No.:77696-421