Order Entry
Puerto Rico
ContactUsLinkComponent
ASTM Method D4815 Ethanol/RFA oxygenate free gasoline 10% Wt, Dr. Ehrenstorfer, LGC Standards
ASTM Method D4815 Ethanol/RFA oxygenate free gasoline 10% Wt, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-798
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
ASTM Method D4815 Ethanol/RFA oxygenate free gasoline 10% Wt, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-798
Supplier:  LGC Standards
Supplier Number:  DRE-GS09000478
New Product
CAS Number:  

Specifications

  • Description:
    ASTM Method D4815 Ethanol/RFA oxygenate free gasoline 10% Wt
  • Size:
    5 x 20 mL
  • Cat. No.:
    77691-798

Specifications

About this item