ASTM Method D7845 Check Standard 0.01-0.05 Wt% in Toluene, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-811
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Toluene
- Description:ASTM Method D7845 Check Standard 0.01-0.05 Wt% in Toluene
- Size:10 x 2 mL
- Cat. No.:77691-811