Michigan Residual Solvents Mixture Kit 699 Non-Inhaled Products 2-5000 µg/mL in Triacetin, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-034
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Triacetin
- Description:Michigan Residual Solvents Mixture Kit 699 Non-Inhaled Products 2-5000 µg/mL in Triacetin
- Size:4 x 1 mL
- Cat. No.:77679-034