Michigan Residual Solvents Mixture 471 1000 µg/mL in Triacetin, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-204
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Triacetin
- Description:Michigan Residual Solvents Mixture 471 1000 µg/mL in Triacetin
- Size:5 x 1 mL
- Cat. No.:77679-204